Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028711.3 | NP_082987.1 | 5 - 6 | 624 | 98 | |
GenBank mRNA | AB106930.1 | - | 5 - 6 | 768 | 98 | |
AK014394.1 | - | 5 - 6 | 625 | 98 | ||
AK021314.1 | - | 6 - 7 | 710 | 98 | ||
AK043831.1 | - | 5 - 6 | 824 | 98 | ||
AK054292.1 | - | 5 - 6 | 1270 | 98 | ||
BC138993.1 | - | 5 - 6 | 631 | 8862015 | 98 | |
BC138994.1 | - | 5 - 6 | 631 | 8862016 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |