Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024134.2 | NP_001019305.1 | 2 - 3 | 549 | 92 | |
NM_001177872.1 | NP_001171343.1 | 2 - 3 | 572 | 92 | ||
GenBank mRNA | AK007445.1 | - | 2 - 3 | 573 | 92 | |
BC027186.1 | - | 2 - 3 | 549 | 4215801 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |