Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172619.3 | NP_766207.2 | 16 - 17 | 2190 | 65 | |
NR_037707.1 | 16 - 17 | 2178 | 65 | |||
NR_037708.1 | 16 - 17 | 2195 | 65 | |||
GenBank mRNA | AF302012.1 | - | 1 - 2 | 67 | 65 | |
AK042525.1 | - | 14 - 15 | 2127 | 65 | ||
AK048600.1 | - | 12 - 13 | 1373 | 65 | ||
AK076334.1 | - | 15 - 16 | 2243 | 65 | ||
AK169468.1 | - | 4 - 5 | 551 | 65 | ||
BC056427.1 | - | 16 - 17 | 2148 | 65 | ||
BC082773.1 | - | 15 - 16 | 2260 | 65 | ||
BC145353.1 | - | 14 - 15 | 2192 | 65 | ||
BC145354.1 | - | 13 - 14 | 1844 | 65 | ||
BC145355.1 | - | 15 - 16 | 2102 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |