Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212442.2 | NP_997607.2 | 1 - 2 | 61 | 68 | |
GenBank mRNA | AF068246.1 | - | 1 - 2 | 49 | 68 | |
AK143946.1 | - | 61 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus 1700 Probe spans exons |