Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300006656.mm.1 |
NM_001284189.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006656.mm.1 |
NM_011982.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006656.mm.1 |
NM_147176.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006656.mm.1 |
TC1300001043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300006653.mm.1 |
NM_001284189.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006653.mm.1 |
NM_011982.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006653.mm.1 |
NM_147176.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006653.mm.1 |
TC1300001043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300006659.mm.1 |
NM_001284189.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006659.mm.1 |
NM_011982.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006659.mm.1 |
NM_147176.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006659.mm.1 |
NM_152134.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006659.mm.1 |
TC1300001043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300006651.mm.1 |
NM_001284189.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006651.mm.1 |
NM_011982.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006651.mm.1 |
NM_147176.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006651.mm.1 |
TC1300001043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300006655.mm.1 |
NM_001284189.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006655.mm.1 |
NM_011982.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006655.mm.1 |
NM_147176.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006655.mm.1 |
TC1300001043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300006657.mm.1 |
NM_001284189.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006657.mm.1 |
NM_011982.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006657.mm.1 |
NM_147176.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006657.mm.1 |
TC1300001043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300006654.mm.1 |
NM_001284189.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006654.mm.1 |
NM_011982.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006654.mm.1 |
NM_147176.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006654.mm.1 |
TC1300001043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300006652.mm.1 |
NM_001284189.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006652.mm.1 |
NM_011982.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006652.mm.1 |
NM_147176.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006652.mm.1 |
TC1300001043.mm.1 |
MTA TC |