Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fut8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045554.1 | 1 - 2 | 516 | 139 | ||
GenBank mRNA | AK133547.1 | - | 1 - 2 | 399 | 139 | |
AK134537.1 | - | 1 - 2 | 190 | 139 | ||
AK157989.1 | - | 1 - 2 | 443 | 139 | ||
AK163494.1 | - | 1 - 2 | 213 | 139 | ||
CJ070536.1 | - | 1 - 2 | 516 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |