Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290500.1 | NP_001277429.1 | 1 - 2 | 251 | 163 | |
NM_025978.4 | NP_080254.2 | 1 - 2 | 251 | 163 | ||
NM_027619.4 | NP_081895.2 | 1 - 2 | 251 | 163 | ||
GenBank mRNA | AK012246.1 | - | 1 - 2 | 213 | 163 | |
AK016619.1 | - | 1 - 2 | 171 | 163 | ||
AK035570.1 | - | 1 - 2 | 221 | 163 | ||
AK041095.1 | - | 1 - 2 | 225 | 164 | ||
AK044266.1 | - | 1 - 2 | 226 | 163 | ||
AK134539.1 | - | 1 - 2 | 240 | 163 | ||
AK163444.1 | - | 1 - 2 | 251 | 163 | ||
AK173315.1 | - | 1 - 2 | 221 | 163 | ||
BC021448.1 | - | 1 - 2 | 259 | 163 | ||
BC024847.1 | - | 1 - 2 | 216 | 5355806 | 163 | |
BC138212.1 | - | 1 - 2 | 213 | 8861231 | 163 | |
BC145944.1 | - | 1 - 2 | 213 | 40131247 | 163 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |