Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200003065.mm.1 |
NM_013780.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003065.mm.1 |
NM_013780.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003065.mm.1 |
TC1200000438.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200003064.mm.1 |
NM_013780.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003064.mm.1 |
NM_013780.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003064.mm.1 |
TC1200000438.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200003066.mm.1 |
NM_013780.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003066.mm.1 |
NM_013780.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003066.mm.1 |
TC1200000438.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200003068.mm.1 |
NM_013780.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003068.mm.1 |
NM_013780.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003068.mm.1 |
TC1200000438.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200003070.mm.1 |
NM_013780.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003070.mm.1 |
NM_013780.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003070.mm.1 |
TC1200000438.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200003071.mm.1 |
NM_013780.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003071.mm.1 |
NM_013780.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003071.mm.1 |
TC1200000438.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200003069.mm.1 |
NM_013780.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003069.mm.1 |
NM_013780.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003069.mm.1 |
TC1200000438.mm.1 |
MTA TC |