Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013024.2 | NP_001013042.1 | 16 - 17 | 2029 | 60 | |
GenBank mRNA | AK135141.1 | - | 16 - 17 | 2081 | 60 | |
BC090999.1 | - | 16 - 17 | 2029 | 6754549 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |