Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013024.2 | NP_001013042.1 | 8 - 9 | 1163 | 88 | |
GenBank mRNA | AK135141.1 | - | 8 - 9 | 1215 | 88 | |
BC090999.1 | - | 8 - 9 | 1163 | 6754549 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |