Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hectd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144788.2 | NP_659037.2 | 36 - 37 | 6843 | 59 | |
GenBank mRNA | AK173106.1 | - | 16 - 17 | 3372 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Mus musculus Inventoried Probe spans exons |