Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271884.1 | NP_001258813.1 | 7 - 8 | 765 | 71 | |
NM_207031.2 | NP_996914.1 | 7 - 8 | 765 | 71 | ||
GenBank mRNA | BC116706.1 | - | 7 - 8 | 731 | 40096570 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |