Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009118.2 | NP_033144.1 | 9 - 10 | 1092 | 71 | |
GenBank mRNA | AK044214.1 | - | 8 - 9 | 773 | 71 | |
AK053278.1 | - | 9 - 10 | 851 | 71 | ||
AK143098.1 | - | 9 - 10 | 851 | 71 | ||
AK149299.1 | - | 9 - 10 | 1101 | 71 | ||
BC016498.1 | - | 9 - 10 | 885 | 4527551 | 71 | |
M24086.1 | - | 9 - 10 | 879 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |