Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163579.1 | NP_001157051.1 | 7 - 8 | 1251 | 155 | |
NM_001163580.1 | NP_001157052.1 | 7 - 8 | 1203 | 155 | ||
NM_028915.3 | NP_083191.2 | 7 - 8 | 1251 | 155 | ||
GenBank mRNA | AK004646.1 | - | 7 - 8 | 1252 | 155 | |
AK016559.1 | - | 7 - 8 | 1190 | 155 | ||
AK029563.1 | - | 7 - 8 | 1168 | 155 | ||
AK173256.1 | - | 7 - 8 | 155 | |||
BC061202.1 | - | 7 - 8 | 1159 | 155 | ||
BC099680.1 | - | 7 - 8 | 1167 | 155 | ||
BC138734.1 | - | 7 - 8 | 1225 | 8861756 | 155 | |
BC138735.1 | - | 7 - 8 | 1225 | 8861757 | 155 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |