Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acad9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172678.3 | NP_766266.3 | 9 - 10 | 1078 | 70 | |
GenBank mRNA | AK028571.1 | - | 8 - 9 | 1792 | 70 | |
AK031820.1 | - | 9 - 10 | 1010 | 70 | ||
AK075984.1 | - | 9 - 10 | 997 | 70 | ||
AK145423.1 | - | 9 - 10 | 986 | 70 | ||
AK206153.1 | - | 3 - 4 | 292 | 70 | ||
BC004737.1 | - | 3 - 4 | 324 | 70 | ||
BC031137.1 | - | 9 - 10 | 1014 | 4971848 | 70 | |
BC032213.1 | - | 9 - 10 | 1019 | 5323723 | 70 | |
BC033277.1 | - | 9 - 10 | 999 | 4241041 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |