Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005508.2 | NP_001005508.2 | 8 - 9 | 1244 | 75 | |
GenBank mRNA | AK031319.1 | - | 8 - 9 | 1015 | 75 | |
AK147757.1 | - | 8 - 9 | 1251 | 75 | ||
AK156351.1 | - | 8 - 9 | 1233 | 75 | ||
AK220204.1 | - | 3 - 4 | 378 | 75 | ||
BC082573.1 | - | 8 - 9 | 1192 | 30548035 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |