Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145991.1 | NP_666103.1 | 9 - 10 | 968 | 86 | |
GenBank mRNA | AK080861.1 | - | 9 - 10 | 1100 | 86 | |
BC027756.1 | - | 9 - 10 | 1069 | 5067496 | 86 | |
BC031127.1 | - | 9 - 10 | 968 | 4972826 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |