Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100025232.mm.1 |
NM_007955.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025232.mm.1 |
NM_007955.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025232.mm.1 |
TC0100003120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100025240.mm.1 |
NM_007955.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025240.mm.1 |
NM_007955.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025240.mm.1 |
TC0100003120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100025235.mm.1 |
NM_007955.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025235.mm.1 |
NM_007955.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025235.mm.1 |
TC0100003120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100025234.mm.1 |
NM_007955.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025234.mm.1 |
NM_007955.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025234.mm.1 |
TC0100003120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100025236.mm.1 |
NM_007955.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025236.mm.1 |
NM_007955.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025236.mm.1 |
TC0100003120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100025231.mm.1 |
NM_007955.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025231.mm.1 |
NM_007955.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025231.mm.1 |
TC0100003120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100025233.mm.1 |
NM_007955.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025233.mm.1 |
NM_007955.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025233.mm.1 |
TC0100003120.mm.1 |
MTA TC |