Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nampt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021524.2 | NP_067499.2 | 5 - 6 | 848 | 139 | |
GenBank mRNA | AF234625.1 | - | 5 - 6 | 708 | 139 | |
AK086415.1 | - | 5 - 6 | 679 | 139 | ||
AK153926.1 | - | 5 - 6 | 679 | 139 | ||
AK166270.1 | - | 5 - 6 | 708 | 139 | ||
AY679720.1 | - | 5 - 6 | 683 | 139 | ||
BC004059.1 | - | 5 - 6 | 656 | 3590087 | 139 | |
BC018358.1 | - | 5 - 6 | 848 | 2609271 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |