Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022312.3 | NP_071707.2 | 4 - 5 | 1588 | 80 | |
GenBank mRNA | AK039390.1 | - | 4 - 5 | 1589 | 80 | |
BC132392.1 | - | 4 - 5 | 1222 | 40130669 | 80 | |
BC138043.1 | - | 4 - 5 | 1222 | 8861062 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |