Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spaca3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029367.1 | NP_083643.1 | 2 - 3 | 399 | 85 | |
GenBank mRNA | AK006357.1 | - | 3 - 4 | 547 | 85 | |
AY572448.1 | - | 2 - 3 | 362 | 85 | ||
AY601763.1 | - | 1 - 2 | 82 | 85 | ||
BC100503.1 | - | 2 - 3 | 399 | 30917230 | 85 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |