Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144827.4 | NP_659076.2 | 11 - 12 | 1562 | 69 | |
GenBank mRNA | BC050788.1 | - | 11 - 12 | 1562 | 6706609 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |