Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Appbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025825.3 | NP_080101.1 | 11 - 12 | 1645 | 80 | |
GenBank mRNA | AK004888.1 | - | 11 - 12 | 1655 | 80 | |
AK005440.1 | - | 11 - 12 | 1351 | 80 | ||
AK122222.1 | - | 1252 | 80 | |||
AK133853.1 | - | 11 - 12 | 1637 | 80 | ||
BC018442.1 | - | 11 - 12 | 1623 | 3157290 | 80 | |
BC019152.1 | - | 11 - 12 | 1646 | 5065139 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |