Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500009259.mm.1 |
NM_001243837.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009259.mm.1 |
NM_001243837.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009259.mm.1 |
TC1500001151.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500009260.mm.1 |
NM_001243837.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009260.mm.1 |
NM_001243837.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009260.mm.1 |
TC1500001151.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500009261.mm.1 |
NM_001243837.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009261.mm.1 |
NM_001243837.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009261.mm.1 |
TC1500001151.mm.1 |
MTA TC |