Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggnbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153144.2 | NP_694784.2 | 11 - 12 | 1704 | 89 | |
GenBank mRNA | AB064543.1 | - | 12 - 13 | 1833 | 89 | |
AY633741.1 | - | 11 - 12 | 1704 | 89 | ||
BC139772.1 | - | 10 - 11 | 1604 | 40111779 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |