Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172147.1 | NP_001165618.1 | 6 - 7 | 1096 | 92 | |
NM_001172148.1 | NP_001165619.1 | 5 - 6 | 1014 | 92 | ||
NM_001290630.1 | NP_001277559.1 | 6 - 7 | 1086 | 92 | ||
NM_153586.2 | NP_705814.1 | 5 - 6 | 1014 | 92 | ||
GenBank mRNA | AK084514.1 | - | 5 - 6 | 949 | 92 | |
AK084629.1 | - | 5 - 6 | 949 | 92 | ||
AK141075.1 | - | 6 - 7 | 1021 | 92 | ||
AK163147.1 | - | 4 - 5 | 934 | 92 | ||
AK164496.1 | - | 6 - 7 | 1044 | 92 | ||
AK166005.1 | - | 5 - 6 | 949 | 92 | ||
BC037024.1 | - | 5 - 6 | 981 | 5368185 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |