Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144811.3 | NP_659060.1 | 1 - 2 | 195 | 69 | |
GenBank mRNA | AB221539.1 | - | 1 - 2 | 69 | 69 | |
AK045714.1 | - | 1 - 2 | 189 | 69 | ||
AY453793.1 | - | 1 - 2 | 185 | 69 | ||
BC021398.1 | - | 1 - 2 | 211 | 5066655 | 69 | |
BC042657.1 | - | 1 - 2 | 317 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |