Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kpnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008379.3 | NP_032405.3 | 19 - 20 | 2654 | 107 | |
GenBank mRNA | AK077709.1 | - | 19 - 20 | 2645 | 107 | |
AK147175.1 | - | 19 - 20 | 2673 | 107 | ||
AK169171.1 | - | 19 - 20 | 2621 | 107 | ||
BC052438.1 | - | 19 - 20 | 2409 | 6405935 | 107 | |
BC052711.1 | - | 19 - 20 | 2442 | 6835239 | 107 | |
BC055115.1 | - | 19 - 20 | 2639 | 6518750 | 107 | |
D45836.1 | - | 19 - 20 | 2545 | 107 | ||
D67015.1 | - | 19 - 20 | 2382 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |