Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc23l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029430.1 | NP_083706.1 | 2 - 3 | 256 | 109 | |
GenBank mRNA | AK015027.1 | - | 2 - 3 | 228 | 109 | |
BC132384.1 | - | 2 - 3 | 256 | 109 | ||
BC145852.1 | - | 2 - 3 | 256 | 40131155 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |