Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033337.4 | NP_001028509.2 | 3 - 4 | 277 | 57 | |
GenBank mRNA | AK078848.1 | - | 3 - 4 | 238 | 57 | |
BC024550.1 | - | 3 - 4 | 227 | 57 | ||
BC030849.1 | - | 3 - 4 | 240 | 57 | ||
BC132621.1 | - | 3 - 4 | 262 | 40130898 | 57 | |
BC132623.1 | - | 3 - 4 | 262 | 40130900 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |