Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Def6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027185.3 | NP_081461.2 | 1 - 2 | 165 | 65 | |
GenBank mRNA | AF329497.1 | - | 1 - 2 | 100 | 65 | |
AK038050.1 | - | 1 - 2 | 136 | 65 | ||
AK088460.1 | - | 1 - 2 | 114 | 65 | ||
AY241695.1 | - | 1 - 2 | 100 | 65 | ||
BC120500.1 | - | 1 - 2 | 165 | 40129423 | 65 | |
BC131953.1 | - | 1 - 2 | 165 | 40130230 | 65 | |
X96705.1 | - | 1 - 2 | 59 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |