Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Def6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027185.3 | NP_081461.2 | 9 - 10 | 1651 | 73 | |
GenBank mRNA | AF329497.1 | - | 9 - 10 | 1586 | 73 | |
AK010356.1 | - | 1 - 2 | 125 | 73 | ||
AK038050.1 | - | 9 - 10 | 1622 | 73 | ||
AK088460.1 | - | 9 - 10 | 1600 | 73 | ||
AY241695.1 | - | 9 - 10 | 1586 | 73 | ||
AY278770.1 | - | 3 - 4 | 467 | 73 | ||
BC120500.1 | - | 9 - 10 | 1651 | 40129423 | 73 | |
BC131953.1 | - | 9 - 10 | 1651 | 40130230 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |