Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039363.2 | NP_001034452.1 | 20 - 21 | 2336 | 61 | |
NM_001286713.1 | NP_001273642.1 | 20 - 21 | 2336 | 61 | ||
NM_001286714.1 | NP_001273643.1 | 20 - 21 | 2331 | 61 | ||
NM_001286716.1 | NP_001273645.1 | 20 - 21 | 2331 | 61 | ||
NM_001286718.1 | NP_001273647.1 | 20 - 21 | 2336 | 61 | ||
NM_001286720.1 | NP_001273649.1 | 20 - 21 | 2336 | 61 | ||
NM_011647.3 | NP_035777.2 | 20 - 21 | 2336 | 61 | ||
GenBank mRNA | AF132986.1 | - | 16 - 17 | 1864 | 61 | |
AK143799.1 | - | 20 - 21 | 2337 | 61 | ||
AK147302.1 | - | 20 - 21 | 2276 | 61 | ||
AK147487.1 | - | 20 - 21 | 2305 | 61 | ||
AK147874.1 | - | 20 - 21 | 2301 | 61 | ||
AK147908.1 | - | 20 - 21 | 2337 | 61 | ||
AK148072.1 | - | 20 - 21 | 2233 | 61 | ||
AK148271.1 | - | 8 - 9 | 908 | 61 | ||
AK170589.1 | - | 20 - 21 | 2327 | 61 | ||
AK171007.1 | - | 20 - 21 | 2305 | 61 | ||
BC052207.1 | - | 6 - 7 | 655 | 61 | ||
BC052449.1 | - | 20 - 21 | 2312 | 6409037 | 61 | |
BC060701.1 | - | 20 - 21 | 2312 | 6409037 | 61 | |
U37775.1 | - | 19 - 20 | 2248 | 61 | ||
U39818.1 | - | 20 - 21 | 2307 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |