Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027231.1 | NP_081507.1 | 3 - 4 | 270 | 99 | |
GenBank mRNA | AK007913.1 | - | 3 - 4 | 285 | 99 | |
AK165600.1 | - | 3 - 4 | 297 | 99 | ||
BC024419.1 | - | 3 - 4 | 270 | 4983359 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |