Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011950.2 | NP_036080.2 | 10 - 11 | 938 | 83 | |
GenBank mRNA | AK146993.1 | - | 10 - 11 | 926 | 83 | |
BC001992.1 | - | 10 - 11 | 907 | 3492903 | 83 | |
U81823.1 | - | 10 - 11 | 895 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |