Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007742.3 | NP_031768.2 | 36 - 37 | 2625 | 99 | |
GenBank mRNA | AK132180.1 | - | 36 - 37 | 2646 | 99 | |
AK159283.1 | - | 36 - 37 | 2643 | 99 | ||
AK159285.1 | - | 36 - 37 | 2646 | 99 | ||
BC042642.1 | - | 29 - 30 | 2195 | 99 | ||
BC050014.1 | - | 36 - 37 | 2625 | 4225038 | 99 | |
M14423.1 | - | 14 - 15 | 975 | 99 | ||
U08020.1 | - | 36 - 37 | 2526 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |