Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lonrf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028894.1 | NP_083170.1 | 1 - 2 | 987 | 102 | |
GenBank mRNA | AK016522.1 | - | 1 - 2 | 987 | 102 | |
AK147751.1 | - | 1 - 2 | 991 | 102 | ||
AK167341.1 | - | 1 - 2 | 1045 | 102 | ||
BC120688.1 | - | 1 - 2 | 889 | 40129611 | 102 | |
BC120690.1 | - | 1 - 2 | 889 | 40129613 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |