Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC052082.1 | - | 3 - 4 | 587 | 6404424 | 68 |
FJ593171.1 | - | 3 - 4 | 620 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |