Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbkbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198100.2 | NP_932768.2 | 7 - 8 | 1118 | 59 | |
GenBank mRNA | AK014173.1 | - | 10 - 11 | 1246 | 59 | |
BC057590.1 | - | 7 - 8 | 1118 | 6810986 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |