Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgcs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291439.1 | NP_001278368.1 | 3 - 4 | 749 | 73 | |
NM_145942.5 | NP_666054.2 | 4 - 5 | 808 | 73 | ||
GenBank mRNA | AK031297.1 | - | 4 - 5 | 697 | 73 | |
AK044835.1 | - | 4 - 5 | 698 | 73 | ||
AK045094.1 | - | 3 - 4 | 637 | 73 | ||
AK078743.1 | - | 3 - 4 | 639 | 73 | ||
AK135551.1 | - | 3 - 4 | 637 | 73 | ||
AK136171.1 | - | 4 - 5 | 697 | 73 | ||
AK145424.1 | - | 3 - 4 | 637 | 73 | ||
AK146042.1 | - | 3 - 4 | 639 | 73 | ||
AK146590.1 | - | 3 - 4 | 639 | 73 | ||
BC023851.1 | - | 4 - 5 | 724 | 5347038 | 73 | |
BC029693.1 | - | 3 - 4 | 654 | 5375374 | 73 | |
BC031363.1 | - | 3 - 4 | 656 | 5357048 | 73 | |
BC034317.1 | - | 2 - 3 | 600 | 5366786 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |