Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erbb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003817.1 | NP_001003817.1 | 5 - 6 | 830 | 86 | |
GenBank mRNA | AK031099.1 | - | 5 - 6 | 698 | 86 | |
AK083669.1 | - | 5 - 6 | 698 | 86 | ||
AK129487.1 | - | 5 - 6 | 800 | 86 | ||
AK142296.1 | - | 5 - 6 | 823 | 86 | ||
BC046811.1 | - | 5 - 6 | 821 | 5702040 | 86 | |
BC053078.1 | - | 5 - 6 | 821 | 5702040 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |