Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500009547.mm.1 |
NM_148938.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009547.mm.1 |
NM_148938.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009547.mm.1 |
TC1500001183.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500009541.mm.1 |
NM_148938.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009541.mm.1 |
NM_148938.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009541.mm.1 |
TC1500001183.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500009542.mm.1 |
NM_148938.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009542.mm.1 |
NM_148938.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009542.mm.1 |
TC1500001183.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500009550.mm.1 |
NM_148938.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009550.mm.1 |
NM_148938.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009550.mm.1 |
TC1500001183.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500009551.mm.1 |
NM_148938.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009551.mm.1 |
NM_148938.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009551.mm.1 |
TC1500001183.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500009549.mm.1 |
NM_148938.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009549.mm.1 |
NM_148938.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009549.mm.1 |
TC1500001183.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500009544.mm.1 |
NM_148938.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009544.mm.1 |
NM_148938.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009544.mm.1 |
TC1500001183.mm.1 |
MTA TC |