Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttll12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183017.2 | NP_898838.2 | 4 - 5 | 740 | 90 | |
GenBank mRNA | AK084879.1 | - | 4 - 5 | 706 | 90 | |
AK134345.1 | - | 4 - 5 | 706 | 90 | ||
AK143496.1 | - | 3 - 4 | 572 | 90 | ||
AK150116.1 | - | 4 - 5 | 706 | 90 | ||
AM690756.1 | - | 4 - 5 | 693 | 90 | ||
BC055368.1 | - | 4 - 5 | 705 | 30055801 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |