Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028821.3 | NP_083097.2 | 5 - 6 | 332 | 137 | |
GenBank mRNA | AK005826.1 | - | 5 - 6 | 418 | 137 | |
BC070454.1 | - | 289 | 137 | |||
BC125392.1 | - | 4 - 5 | 314 | 40129907 | 137 | |
BC125394.1 | - | 4 - 5 | 310 | 40129909 | 137 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |