Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199198.2 | NP_954668.2 | 9 - 10 | 1068 | 78 | |
NR_028447.1 | 9 - 10 | 1079 | 78 | |||
NR_028448.1 | 8 - 9 | 999 | 78 | |||
NR_028449.1 | 9 - 10 | 1079 | 78 | |||
GenBank mRNA | AK040235.1 | - | 9 - 10 | 1031 | 78 | |
AK045028.1 | - | 8 - 9 | 1279 | 78 | ||
AK154045.1 | - | 8 - 9 | 963 | 78 | ||
BC013700.1 | - | 9 - 10 | 1032 | 78 | ||
BC064018.1 | - | 9 - 10 | 995 | 3980699 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |