Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Creld2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029720.2 | NP_083996.1 | 5 - 6 | 651 | 95 | |
GenBank mRNA | AK017880.1 | - | 5 - 6 | 652 | 95 | |
AK165884.1 | - | 5 - 6 | 650 | 95 | ||
AK198081.1 | - | 1 - 2 | 127 | 95 | ||
BC047370.1 | - | 5 - 6 | 602 | 4209333 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |