Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdfy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175546.4 | NP_780755.2 | 9 - 10 | 1163 | 70 | |
GenBank mRNA | AK045072.1 | - | 8 - 9 | 894 | 70 | |
AK086275.1 | - | 9 - 10 | 1176 | 70 | ||
AK151817.1 | - | 9 - 10 | 1119 | 70 | ||
BC116632.1 | - | 9 - 10 | 1109 | 40056912 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |