Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itm2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008410.2 | NP_032436.1 | 2 - 3 | 391 | 124 | |
GenBank mRNA | AB030203.1 | - | 2 - 3 | 411 | 124 | |
AB030204.1 | - | 1 - 2 | 207 | 124 | ||
AK002599.1 | - | 2 - 3 | 354 | 124 | ||
AK003224.1 | - | 2 - 3 | 358 | 124 | ||
AK003587.1 | - | 2 - 3 | 380 | 124 | ||
AK003928.1 | - | 2 - 3 | 379 | 124 | ||
AK005125.1 | - | 2 - 3 | 382 | 124 | ||
AK076139.1 | - | 2 - 3 | 400 | 124 | ||
AK077482.1 | - | 2 - 3 | 393 | 124 | ||
AK088270.1 | - | 2 - 3 | 379 | 124 | ||
AK150378.1 | - | 2 - 3 | 411 | 124 | ||
AK150396.1 | - | 2 - 3 | 410 | 124 | ||
AK151714.1 | - | 2 - 3 | 410 | 124 | ||
AK151889.1 | - | 2 - 3 | 411 | 124 | ||
AK152068.1 | - | 2 - 3 | 399 | 124 | ||
AK152516.1 | - | 2 - 3 | 411 | 124 | ||
AK152650.1 | - | 2 - 3 | 411 | 124 | ||
AK152731.1 | - | 2 - 3 | 382 | 124 | ||
AK152983.1 | - | 2 - 3 | 403 | 124 | ||
AK153103.1 | - | 2 - 3 | 354 | 124 | ||
AK159307.1 | - | 2 - 3 | 354 | 124 | ||
AK159340.1 | - | 2 - 3 | 379 | 124 | ||
AK159378.1 | - | 2 - 3 | 410 | 124 | ||
AK159426.1 | - | 2 - 3 | 379 | 124 | ||
AK159619.1 | - | 2 - 3 | 403 | 124 | ||
AK159628.1 | - | 2 - 3 | 379 | 124 | ||
AK159792.1 | - | 2 - 3 | 379 | 124 | ||
AK159962.1 | - | 2 - 3 | 382 | 124 | ||
AK160101.1 | - | 2 - 3 | 379 | 124 | ||
AK160725.1 | - | 2 - 3 | 410 | 124 | ||
BC004731.1 | - | 2 - 3 | 362 | 3583154 | 124 | |
BC010320.1 | - | 2 - 3 | 394 | 3599190 | 124 | |
BC021786.1 | - | 2 - 3 | 400 | 4458918 | 124 | |
U76253.1 | - | 2 - 3 | 391 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |