Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026083.2 | NP_080359.2 | 12 - 13 | 2996 | 73 | |
GenBank mRNA | AK011176.1 | - | 1 - 2 | 268 | 73 | |
BC082576.1 | - | 1 - 2 | 507 | 73 | ||
BC094575.1 | - | 5 - 6 | 1542 | 73 | ||
BC138264.1 | - | 12 - 13 | 2945 | 8861283 | 73 | |
CF742012.1 | - | 1 - 2 | 507 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |