Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK007218.1 | - | 2 - 3 | 340 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |